Skip to Content
MilliporeSigma
Benzyldimethyltetradecylammonium chloride dihydrate 98%

Benzyldimethyltetradecylammonium chloride dihydrate

Benzyldimethyltetradecylammonium chloride dihydrate 98%
Linear Formula:
CH3(CH2)13N(Cl)(CH3)2CH2C6H5 · 2H2O
CAS Number:
Molecular Weight:
404.07

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing