Skip to Content
MilliporeSigma
Benzotriazole analytical standard

Benzotriazole

Benzotriazole analytical standard
Synonym(s):
1,2,3-Benzotriazole, 1H-Benzotriazole
Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing