Skip to Content
MilliporeSigma
Benzalkonium chloride ≥95.0% ((calculated on dry substance), T)

Benzalkonium chloride

Benzalkonium chloride ≥95.0% ((calculated on dry substance), T)
Synonym(s):
Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride
Linear Formula:
C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
CAS Number:
EC Number:

Products

Product Category
Brand
Physical Form
Product Line
Purity
Agency
Chemical Composition
Quality Segment
Application
Showing 1-3 of 3
Compare
Product Number
Description
Pricing