Skip to Content
MilliporeSigma
Behenic acid analytical standard

Behenic acid

Behenic acid analytical standard
Synonym(s):
Docosanoic acid
Linear Formula:
CH3(CH2)20COOH
CAS Number:
Molecular Weight:
340.58
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing