Skip to Content
MilliporeSigma
β-Cyclodextrin hydrate 99%

β-Cyclodextrin hydrate

β-Cyclodextrin hydrate 99%
Synonym(s):
β-Schardinger dextrin, Cycloheptaamylose, Schardinger β-Dextrin hydrate
Empirical Formula (Hill Notation):
C42H70O35 · xH2O
CAS Number:
Molecular Weight:
1134.98 (anhydrous basis)
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing