Skip to Content
MilliporeSigma
B-Chlorocatecholborane 97%

B-Chlorocatecholborane

B-Chlorocatecholborane 97%
Synonym(s):
2-Chloro-1,3,2-benzodioxaborole
Empirical Formula (Hill Notation):
C6H4BClO2
CAS Number:
Molecular Weight:
154.36

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing