![Baicalin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/152/188/73fc86ea-cbeb-4e35-ad5b-38640a544e6e/640/73fc86ea-cbeb-4e35-ad5b-38640a544e6e.png)
Baicalin
![Baicalin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/152/188/73fc86ea-cbeb-4e35-ad5b-38640a544e6e/640/73fc86ea-cbeb-4e35-ad5b-38640a544e6e.png)
Synonym(s):
5,6-Dihydroxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, Baicalein 7-O-β-D-glucuronide
Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing