Skip to Content
MilliporeSigma
Azetidine hydrochloride 97%

Azetidine hydrochloride

Azetidine hydrochloride 97%
Synonym(s):
Trimethyleneimine hydrochloride
Empirical Formula (Hill Notation):
C3H7N · HCl
CAS Number:
Molecular Weight:
93.56

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing