Skip to Content
MilliporeSigma
Azaserine Hybri-Max™, γ-irradiated, 50x, lyophilized powder, BioXtra, suitable for hybridoma

Azaserine

Azaserine Hybri-Max™, γ-irradiated, 50x, lyophilized powder, BioXtra, suitable for hybridoma
Synonym(s):
O-Diazoacetyl-L-serine
Empirical Formula (Hill Notation):
C5H7N3O4
CAS Number:
Molecular Weight:
173.13
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Mode of Action
Optical Isomer
Amino Acid
Sterilization Method
Technique
Showing 1-2 of 2