Skip to Content
MilliporeSigma
Azaerythromycin A certified reference material, pharmaceutical secondary standard

Azaerythromycin A

Azaerythromycin A certified reference material, pharmaceutical secondary standard
Synonym(s):
9-Deoxo-9a-aza-9a-homoerythromycin A, Azaerythromycin A
Empirical Formula (Hill Notation):
C37H70N2O12
CAS Number:
Molecular Weight:
734.96

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade