Skip to Content
MilliporeSigma
Avobenzone analytical standard

Avobenzone

Avobenzone analytical standard
Synonym(s):
1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione, Butyl methoxydibenzoylmethane
Empirical Formula (Hill Notation):
C20H22O3
CAS Number:
Molecular Weight:
310.39
EC Number:

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique