Skip to Content
MilliporeSigma
Atovaquone Related Compound A certified reference material, pharmaceutical secondary standard

Atovaquone Related Compound A

Atovaquone Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
cis-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthoquinone
Empirical Formula (Hill Notation):
C22H19ClO3
CAS Number:
Molecular Weight:
366.84

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade