Skip to Content
MilliporeSigma
Atomoxetine Related Compound A United States Pharmacopeia (USP) Reference Standard

Atomoxetine Related Compound A

Atomoxetine Related Compound A United States Pharmacopeia (USP) Reference Standard
Synonym(s):
α-[2-(Methylamino)ethyl]benzyl alcohol, 3-(Methylamino)-1-phenylpropan-1-ol
Linear Formula:
C6H5CH(CH2CH2NHCH3)OH
CAS Number:
Molecular Weight:
165.23

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade