Skip to Content
MilliporeSigma
ART® low retention SoftFit L Reload System™ maximum volume 300 μL, Tip design ART 300L, Reload insert

ART® low retention SoftFit L Reload System

ART® low retention SoftFit L Reload System™ maximum volume 300 μL, Tip design ART 300L, Reload insert

Products

Product Category
Manufacturer
Volume (µL)
Color
Package Size
Sterility
Technique
Showing 1-20 of 34
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2