Skip to Content
MilliporeSigma
Arachidonoyl coenzyme A lithium salt ≥85%

Arachidonoyl coenzyme A lithium salt

Arachidonoyl coenzyme A lithium salt ≥85%
Synonym(s):
cis,cis,cis,cis-5,8,11,14-Eicosatetraenoyl coenzyme A lithium salt
Empirical Formula (Hill Notation):
C41H66N7O17P3S · xLi+
CAS Number:
Molecular Weight:
1053.99 (free acid basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing