Skip to Content
MilliporeSigma
Anti-USP10 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-USP10 antibody produced in rabbit

Anti-USP10 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Deubiquitinating enzyme 10 antibody produced in rabbit, Anti-Ubiquitin carboxyl-terminal hydrolase 10 antibody produced in rabbit, Anti-Ubiquitin thioesterase 10 antibody produced in rabbit, Anti-Ubiquitin-specific-processing protease 10 antibody produced in rabbit, Anti-UBPO, Anti-Ubiquitin specific peptidase 10, Anti-de-ubiquitinating enzyme 10, Anti-ubiquitin carboxyl-terminal hydrolase 10

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique