Skip to Content
MilliporeSigma
Anti-RPS6KA6 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-RPS6KA6 antibody produced in rabbit

Anti-RPS6KA6 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-90 kDa ribosomal protein S6 kinase 6 antibody produced in rabbit, Anti-RSK-4 antibody produced in rabbit, Anti-Ribosomal S6 kinase 4 antibody produced in rabbit, Anti-Ribosomal protein S6 kinase α-6 antibody produced in rabbit, Anti-S6K-α 6 antibody produced in rabbit, Anti-p90-RSK 6 antibody produced in rabbit, Anti-pp90RSK4 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique