Skip to Content
MilliporeSigma
Anti-PABPC5 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-PABPC5 antibody produced in rabbit

Anti-PABPC5 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-PABP 5 antibody produced in rabbit, Anti-Poly(A)-binding protein 5 antibody produced in rabbit, Anti-Polyadenylate-binding protein 5 antibody produced in rabbit, PABP 5, PABP5, Poly(A)-binding protein 5, Polyadenylate-binding protein 5

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique