Skip to Content
MilliporeSigma
Anti-MYOM2 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-MYOM2 antibody produced in rabbit

Anti-MYOM2 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
165 kDa connectin-associated protein, 165 kDa titin-associated protein, M-protein, Myomesin family member 2, Myomesin-2, Anti-165 kDa connectin-associated protein antibody produced in rabbit, Anti-165 kDa titin-associated protein antibody produced in rabbit, Anti-M-protein antibody produced in rabbit, Anti-Myomesin-2 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique