Skip to Content
MilliporeSigma
Anti-LITAF antibody produced in rabbit

Anti-LITAF antibody produced in rabbit

Anti-LITAF antibody produced in rabbit
Synonym(s):
Anti-LPS-induced TNF-α factor antibody produced in rabbit, Anti-Lipopolysaccharide-induced tumor necrosis factor-α factor antibody produced in rabbit, Anti-Small integral membrane protein of lysosome/late endosome antibody produced in rabbit, Anti-p53-induced protein 7 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique