Anti-KRT9 antibody produced in rabbit
Synonym(s):
Anti-CK-9 antibody produced in rabbit, Anti-Cytokeratin-9 antibody produced in rabbit, Anti-K9 antibody produced in rabbit, Anti-Keratin, type I cytoskeletal 9 antibody produced in rabbit, Anti-Keratin-9 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1