Skip to Content
MilliporeSigma
Anti-JPH1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-JPH1 antibody produced in rabbit

Anti-JPH1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-JP-1 antibody produced in rabbit, Anti-Junctophilin type 1 antibody produced in rabbit, Anti-Junctophilin-1 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique