Skip to Content
MilliporeSigma
Anti-HSV antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Anti-HSV antibody produced in rabbit

Anti-HSV antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Herpes Simplex Virus

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1