![Anti-HNRNPH2 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/198/085/1c96a9bf-fc42-4262-9945-ec24bc7546a2/640/1c96a9bf-fc42-4262-9945-ec24bc7546a2.jpg)
Anti-HNRNPH2 antibody produced in rabbit
![Anti-HNRNPH2 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/198/085/1c96a9bf-fc42-4262-9945-ec24bc7546a2/640/1c96a9bf-fc42-4262-9945-ec24bc7546a2.jpg)
Synonym(s):
Anti-FTP-3, Anti-HNRNPH2, Anti-Heterogeneous nuclear ribonucleoprotein H′, Anti-hnRNP H′
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing