![Anti-GRHL1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/104/269/76751c4b-50f8-449a-9e8a-b182b0c020e4/640/76751c4b-50f8-449a-9e8a-b182b0c020e4.jpg)
Anti-GRHL1 antibody produced in rabbit
![Anti-GRHL1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/104/269/76751c4b-50f8-449a-9e8a-b182b0c020e4/640/76751c4b-50f8-449a-9e8a-b182b0c020e4.jpg)
Synonym(s):
Anti-Grainyhead-like protein 1 homolog antibody produced in rabbit, Anti-Mammalian grainyhead antibody produced in rabbit, Anti-NH32 antibody produced in rabbit, Anti-Transcription factor CP2-like 2 antibody produced in rabbit, Anti-Transcription factor LBP-32 antibody produced in rabbit, Anti-Grainyhead-like 1 (Drosophila), Anti-LBP-32, Anti-LBP32, Anti-MGR, Anti-TFCP2L2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing