Skip to Content
MilliporeSigma
Anti-GIT1 antibody produced in rabbit

Anti-GIT1 antibody produced in rabbit

Anti-GIT1 antibody produced in rabbit
Synonym(s):
Anti-ARF GTPase-activating protein GIT1 antibody produced in rabbit, Anti-Cat-1 antibody produced in rabbit, Anti-Cool-associated and tyrosine-phosphorylated protein 1 antibody produced in rabbit, Anti-G protein-coupled receptor kinase-interactor 1 antibody produced in rabbit, Anti-GRK-interacting protein 1 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique