Skip to Content
MilliporeSigma
anti-Diphenylglyoxime 97%

anti-Diphenylglyoxime

anti-Diphenylglyoxime 97%
Synonym(s):
α-Benzil dioxime
Linear Formula:
C6H5C(=NOH)C(=NOH)C6H5
CAS Number:
Molecular Weight:
240.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing