Skip to Content
MilliporeSigma
vibrant-m

Antibiotic Antimycotic Solution (100×), Stabilized

vibrant-m

Products

Product Category
Brand
Color
Physical Form
Product Line
Activity Spectrum
Quality Segment
Application
Mode of Action
Sterilization Method
Technique
Showing 1-1 of 1