Skip to Content
MilliporeSigma
Anti-β-Amyloid antibody, Mouse monoclonal clone BAM-10, ascites fluid

Anti-β-Amyloid antibody, Mouse monoclonal

Anti-β-Amyloid antibody, Mouse monoclonal clone BAM-10, ascites fluid
Synonym(s):
Anti-A-BETA, Anti-Amyloid β Precursor Protein, Clone BAM91

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing