Skip to Content
MilliporeSigma
Anti-Aurora-A Kinase antibody, Mouse monoclonal clone 35C1, purified from hybridoma cell culture

Anti-Aurora-A Kinase antibody, Mouse monoclonal

Anti-Aurora-A Kinase antibody, Mouse monoclonal clone 35C1, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Aurora-A Kinase, Anti-ARK-1, Anti-AURORA2, Anti-Aurora-related kinase 1, Anti-BTAK, Anti-STK15, Anti-STK6, Anti-Serine/threonine protein kinase 15

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1