Skip to Content
MilliporeSigma
Amylamine ≥99%, FG

Amylamine

Amylamine ≥99%, FG
Synonym(s):
Pentylamine, 1-Aminopentane, n-Amylamine
Linear Formula:
CH3(CH2)4NH2
CAS Number:
Molecular Weight:
87.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing