Skip to Content
MilliporeSigma
Ammonium Sulfate, Molecular Biology Grade Absorbance (260 nm and 280 nm): ≤0.01.

Ammonium Sulfate, Molecular Biology Grade

Ammonium Sulfate, Molecular Biology Grade Absorbance (260 nm and 280 nm): ≤0.01.
Synonym(s):
Ammonium sulfate, Mascagnite
Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1