Skip to Content
MilliporeSigma
Ammonium diethyldithiocarbamate

Ammonium diethyldithiocarbamate

Ammonium diethyldithiocarbamate
Synonym(s):
Diethyldithiocarbamic acid ammonium salt
Linear Formula:
(C2H5)2NC(S)SSNH4
CAS Number:
Molecular Weight:
166.31
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing