Skip to Content
MilliporeSigma
Ammonium-15N2 sulfate 30 atom % 15N

Ammonium-15N2 sulfate

Ammonium-15N2 sulfate 30 atom % 15N
Synonym(s):
15N Labeled ammonium sulfate, Diazanium-15N2 sulfate
Linear Formula:
(15NH4)2SO4
CAS Number:
Molecular Weight:
134.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Application
Markush Class
Markush Group
Technique