Skip to Content
MilliporeSigma
α-Methylene-γ-butyrolactone 97%

α-Methylene-γ-butyrolactone

α-Methylene-γ-butyrolactone 97%
Synonym(s):
3-Methylenedihydro-2(3H)-furanone, Tulipane
Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing