Skip to Content
MilliporeSigma
α-Methyl-DL-phenylalanine 98%

α-Methyl-DL-phenylalanine

α-Methyl-DL-phenylalanine 98%
Synonym(s):
(±)-2-Amino-2-methyl-3-phenylpropionic acid
Linear Formula:
C6H5CH2C(CH3)(NH2)CO2H
CAS Number:
Molecular Weight:
179.22

Products

Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing