Skip to Content
MilliporeSigma
α-Melanocyte stimulating hormone ≥97% (HPLC)

α-Melanocyte stimulating hormone

α-Melanocyte stimulating hormone ≥97% (HPLC)
Synonym(s):
α-MSH, α-Melanotropin, α-Melanocyte-Stimulating Hormone
Empirical Formula (Hill Notation):
C77H109N21O19S
CAS Number:
Molecular Weight:
1664.88

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility