Skip to Content
MilliporeSigma
Amantadine hydrochloride British Pharmacopoeia (BP) Reference Standard

Amantadine hydrochloride

Amantadine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1-Adamantanamine hydrochloride, 1-Adamantylamine hydrochloride, 1-Aminoadamantane hydrochloride, NSC 83653, Tricyclo[3.3.1.13,7]decan-1-amine hydrochloride, Amantadine hydrochloride
Empirical Formula (Hill Notation):
C10H17N · HCl
CAS Number:
Molecular Weight:
187.71

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade