Skip to Content
MilliporeSigma
Aluminum acetylacetonate purified by sublimation, 99.999% trace metals basis

Aluminum acetylacetonate

Aluminum acetylacetonate purified by sublimation, 99.999% trace metals basis
Synonym(s):
Al(acac)3, Aluminum 2,4-pentanedionate
Linear Formula:
Al(C5H7O2)3
CAS Number:
Molecular Weight:
324.31
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-2 of 2