Skip to Content
MilliporeSigma
Alpha-Tricalcium phosphate (TCP), powder, ≥98%, <500 μm particle size

Alpha-Tricalcium phosphate

Alpha-Tricalcium phosphate (TCP), powder, ≥98%, <500 μm particle size
Synonym(s):
α-Tri-Calcium phosphate, α-TCP, α-Tricalcium phosphate
Linear Formula:
Ca3(PO4)2
CAS Number:
Molecular Weight:
310.18

Products

Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing