Skip to Content
MilliporeSigma
Alpha Cyclodextrin United States Pharmacopeia (USP) Reference Standard

Alpha Cyclodextrin

Alpha Cyclodextrin United States Pharmacopeia (USP) Reference Standard
Synonym(s):
α-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose
Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing