Alloxan monohydrate
Synonym(s):
2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil
Empirical Formula (Hill Notation):
C4H2N2O4 · H2O
CAS Number:
Molecular Weight:
160.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing