Skip to Content
MilliporeSigma
Albendazole sulfoxide ≥97% (HPLC)

Albendazole sulfoxide

Albendazole sulfoxide ≥97% (HPLC)
Synonym(s):
Ricobendazole, Albendazole oxide, Albendazole sulfoxide, Methyl [5-(propane-1-sulfinyl)-1H-benzoimidazol-2-yl]-carbamate
Empirical Formula (Hill Notation):
C12H15N3O3S
CAS Number:
Molecular Weight:
281.33

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing