Skip to Content
MilliporeSigma
Ala-Pro hydrate

Ala-Pro hydrate

Ala-Pro hydrate
Synonym(s):
L-Alanyl-L-proline hydrate
Empirical Formula (Hill Notation):
C8H14N2O3 · xH2O
CAS Number:
Molecular Weight:
186.21 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing