Skip to Content
MilliporeSigma
Ala-Gln ≥98% (HPLC)

Ala-Gln

Ala-Gln ≥98% (HPLC)
Synonym(s):
Alanyl-glutamine, Glutamine-S
Empirical Formula (Hill Notation):
C8H15N3O4
CAS Number:
Molecular Weight:
217.22

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Sterilization Method
Technique