Skip to Content
MilliporeSigma
Agnuside United States Pharmacopeia (USP) Reference Standard

Agnuside

Agnuside United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(1S,4aR,5S,7aS)-1,4a,5,7a-Tetrahydro-5-hydroxy-7-[[(4-hydroxybenzoyl)oxy]methyl]cyclopenta[c]pyran-1-yl β-D-glucopyranoside, Buddlejoside A
Empirical Formula (Hill Notation):
C22H26O11
CAS Number:
Molecular Weight:
466.44

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3