![α-D-Galactose 1-phosphate dipotassium salt pentahydrate Type II, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/328/827/ca3ef601-1cfc-4c0a-be4f-1c076b0a6334/640/ca3ef601-1cfc-4c0a-be4f-1c076b0a6334.png)
α-D-Galactose 1-phosphate dipotassium salt pentahydrate
![α-D-Galactose 1-phosphate dipotassium salt pentahydrate Type II, ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/328/827/ca3ef601-1cfc-4c0a-be4f-1c076b0a6334/640/ca3ef601-1cfc-4c0a-be4f-1c076b0a6334.png)
Synonym(s):
α-D-Galactopyranosyl 1-phosphate dipotassium salt, α-D-Galactose-1-phosphate, Dipotassium Salt, Pentahydrate
Empirical Formula (Hill Notation):
C6H11K2O9P · 5H2O
CAS Number:
Molecular Weight:
426.39
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing