Skip to Content
MilliporeSigma
Adenosine 5′-triphosphate di(tris) salt hydrate ≥95%

Adenosine 5′-triphosphate di(tris) salt hydrate

Adenosine 5′-triphosphate di(tris) salt hydrate ≥95%
Synonym(s):
5′-ATP-DiTris
Empirical Formula (Hill Notation):
C10H16N5O13P3 · 2C4H11NO3 · xH2O
CAS Number:
Molecular Weight:
749.45 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing