Skip to Content
MilliporeSigma
Adenosine 5′-triphosphate dipotassium salt hydrate from microbial, ≥92% (HPLC)

Adenosine 5′-triphosphate dipotassium salt hydrate

Adenosine 5′-triphosphate dipotassium salt hydrate from microbial, ≥92% (HPLC)
Synonym(s):
5′-ATP-K2
Empirical Formula (Hill Notation):
C10H14K2N5O13P3 · xH2O
CAS Number:
Molecular Weight:
583.36 (anhydrous basis)

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1