Skip to Content
MilliporeSigma
Adenosine 5′-phosphosulfate sodium salt ≥85%

Adenosine 5′-phosphosulfate sodium salt

Adenosine 5′-phosphosulfate sodium salt ≥85%
Synonym(s):
APS
Empirical Formula (Hill Notation):
C10H14N5O10PS
CAS Number:
Molecular Weight:
427.28

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing